missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy Rosiglitazone Beta-D-Glucuronide (mixture of diastereomers), TRC

Product Code. 30405392
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405392 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405392 Supplier Toronto Research Chemicals Supplier No. TRCH95344510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxy Rosiglitazone beta-D-Glucuronide
CAS 288853-59-8
Molecular Formula C24 H27 N3 O10 S
Quantity 10 mg
Synonym 5-Hydroxy Rosiglitazone-B-D-Glucuronide
InChI Formula InChI=1S/C24H27N3O10S/c1-27(8-9-35-13-4-2-12(3-5-13)10-15-21(31)26-24(34)38-15)16-7-6-14(11-25-16)36-23-19(30)17(28)18(29)20(37-23)22(32)33/h2-7,11,15,17-20,23,28-30H,8-10H2,1H3,(H,32,33)(H,26,31,34)/t15?,17-,18-,19+,20-,23+/m0/s1
SMILES CN(CCOc1ccc(CC2SC(=O)NC2=O)cc1)c3ccc(O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O)cn3
IUPAC Name (2S,3S,4S,5R,6S)-6-[6-[2-[4-[(2,4-dioxo-1,3-thiazolidin-5-yl)methyl]phenoxy]ethyl-methylamino]pyridin-3-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 549.55
Formula Weight 549.1417
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.