missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-2-carboxylic acid, 97%

Product Code. 11360214
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360214 1 g 1g
11370214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360214 Supplier Thermo Scientific Alfa Aesar Supplier No. L00786.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21598-06-1 | C9H6NO3 | 176.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21598-06-1
Molecular Formula C9H6NO3
Molecular Weight (g/mol) 176.15
MDL Number MFCD00005615
InChI Key BIMHWDJKNOMNLD-UHFFFAOYSA-M
Synonym 5-hydroxyindole-2-carboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxyindole-2-carboxylicacid, zlchem 754, pubchem1716, acmc-20am33, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxy-1h-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-hydroxy
PubChem CID 88958
IUPAC Name 5-hydroxy-1H-indole-2-carboxylic acid
SMILES OC1=CC=C2NC(=CC2=C1)C([O-])=O

Specifications

Melting Point ∽245°C (decomposition)
Quantity 1 g
Beilstein 153214
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-Hydroxyindole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.