missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxymethyl-2-furaldehyde, 97%

Product Code. 11474084
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474084 25 g 25g
11454084 1 g 1g
11464084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474084 Supplier Thermo Scientific Alfa Aesar Supplier No. A12475.14
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-47-0 | C6H6O3 | 126.111 g/mol

5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5'-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5′-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.

Solubility
Soluble in water, alcohol, ethyl acetate, acetone, dimethylformamide, benzene, ether and chloroform.

Notes
Air and light sensitive. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-47-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00003234
InChI Key NOEGNKMFWQHSLB-UHFFFAOYSA-N
Synonym 5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
PubChem CID 237332
ChEBI CHEBI:412516
IUPAC Name 5-(hydroxymethyl)furan-2-carbaldehyde
SMILES C1=C(OC(=C1)C=O)CO

Specifications

Melting Point 32°C to 35°C
Density 1.206
Boiling Point 114°C to 116°C (1 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.562
Quantity 25 g
Beilstein 110889
Sensitivity Air and light sensitive
Merck Index 14,4832
Solubility Information Soluble in water,alcohol,ethyl acetate,acetone,dimethylformamide,benzene,ether and chloroform.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 5-Hydroxymethyl-2-furaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.