missing translation for 'onlineSavingsMsg'
Learn More

5’-Hydroxymethyl 5’-Desmethyl Meloxicam, TRC

Product Code. 30011827
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011827 2.5 mg 2.5mg
30006021 10 mg 10mg
30004200 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011827 Supplier Toronto Research Chemicals Supplier No. TRCH9478102.5MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130262-92-9
Molecular Formula C14 H13 N3 O5 S2
Molecular Weight (g/mol) 367.4
Synonym 2H-1,2-Benzothiazine-3-carboxamide, 4-hydroxy-N-[5-(hydroxymethyl)-2-thiazolyl]-2-methyl-, 1,1-dioxide, 4-Hydroxy-N-[5-(hydroxymethyl)-2-thiazolyl]-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, 5-Hydroxy ethyl meloxicam, 5'-Hydroxymethyl 5'-Desmethyl Meloxicam
IUPAC Name 4,5-dihydroxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
SMILES CN1C(=C(O)c2c(O)cccc2S1(=O)=O)C(=O)Nc3ncc(C)s3

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C14H13N3O5S2/c1-7-6-15-14(23-7)16-13(20)11-12(19)10-8(18)4-3-5-9(10)24(21,22)17(11)2/h3-6,18-19H,1-2H3,(H,15,16,20)
Formula Weight 367.0297
Percent Purity >95%
Chemical Name or Material 5'-Hydroxymethyl 5'-Desmethyl Meloxicam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.