missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)furfural, 98%

Product Code. 10654972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10654972 1 g Glass bottle 1g
10316520 5 g Glass bottle 5g
11986161 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10654972 Supplier Thermo Scientific Acros Supplier No. 121460010
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-47-0 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-47-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
InChI Key NOEGNKMFWQHSLB-UHFFFAOYSA-N
Synonym 5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
PubChem CID 237332
ChEBI CHEBI:412516
IUPAC Name 5-(hydroxymethyl)furan-2-carbaldehyde
SMILES C1=C(OC(=C1)C=O)CO

Specifications

Melting Point 30.0°C to 34.0°C
Color Yellow
Boiling Point 114°C to 116°C (1.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5627
Quantity 1 g
Beilstein 18, V,1, 130
Merck Index 15,4871
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 126.11
Percent Purity 98%
Physical Form Crystalline Powder, Chunks or Liquid
Chemical Name or Material 5-(Hydroxymethyl)furfural
Show More Show Less
compliance-icons
Product Identifier
  • 5-(Hydroxymethyl)furfural
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.