missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxymethyluracil, TRC

Product Code. 30391273
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391273 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391273 Supplier Toronto Research Chemicals Supplier No. TRCH9483751G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4433-40-3
Molecular Formula C5 H6 N2 O3
Quantity 1 g
Synonym 2,4(1H,3H)-Pyrimidinedione, 5-(hydroxymethyl)- (9CI, ACI), 5-(Hydroxymethyl)-2,4(1H,3H)-pyrimidinedione (ACI), Uracil, 5-(hydroxymethyl)- (6CI, 7CI, 8CI), 5-(Hydroxymethyl)-1,3-dihydropyrimidine-2,4-dione, 5-(Hydroxymethyl)pyrimidine-2,4-diol, 5-(Hydroxymethyl)uracil, 5-Hydroxymethyl-1H-pyrimidine-2,4-dione, NSC 20901, α-Hydroxythymine, 5-Hydroxymethyluracil
InChI Formula InChI=1S/C5H6N2O3/c8-2-3-1-6-5(10)7-4(3)9/h1,8H,2H2,(H2,6,7,9,10)
SMILES OCc1cnc(O)nc1O
IUPAC Name 5-(hydroxymethyl)pyrimidine-2,4-diol
Molecular Weight (g/mol) 142.11
Formula Weight 142.0378
Chemical Name or Material 5-Hydroxymethyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.