missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyquinoline, TRC

Product Code. 30405691
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405691 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405691 Supplier Toronto Research Chemicals Supplier No. TRCH939950250MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 578-67-6
Molecular Formula C9H7NO
Quantity 250 mg
Synonym 5-Quinolinol, 5-Hydroxyquinoline, NSC 405729
InChI Formula InChI=1S/C9H7NO/c11-9-5-1-4-8-7(9)3-2-6-10-8/h1-6,11H
SMILES Oc1cccc2ncccc12
IUPAC Name quinolin-5-ol
Molecular Weight (g/mol) 145.16
Formula Weight 145.0528
Chemical Name or Material 5-Hydroxyquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.