missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5-IAF (5-iodoacetamido-fluorescein)

Product Code. 10649148
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10649148 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10649148 Supplier Thermo Scientific™ Supplier No. 62246
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-IAF (5-iodoacetamido-fluorescein)

Thermo Scientific 5-Iodoacetamido-Fluorescein (5-IAF) is a high-performance derivative of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes. 5-Iodoacetamido-Fluorescein is a sulfhydryl-reactive derivative of fluorescein that labels proteins and other molecules having free thiols (cysteine side chains)

Properties of 5-Iodoacetamido-Fluorescein:
  • Alternative names: 5-IAF, 5-iodoacetamidofluorescein
  • Chemical name: Acetamide, N-(3',6'-dihydroxy-3-oxospiro (isobenzofuran-1(3H), 9'-(9H)xanthen)-5-yl)-2-iodo
  • Molecular weight: 515.26±3
  • Excitation source: 488 nm spectral line, argon-ion laser
  • Excitation wavelength: 494 nm
  • Emission wavelength: 518 nm
  • Extinction coefficient: > 80,000 M-1cm-1
  • CAS #: 63368-54-7
  • Solubility: Soluble in DMF; aqueous buffers at pH > 6
  • Reactive groups: Iodoacetamide, reacts with sulfhydryls at pH 7.0 to 7.5

    Applications:
  • Label antibodies for use as immunofluorescent probes
  • Label oligonucleotides for hybridization probes
  • Detect proteins in gels and on Western blots
TRUSTED_SUSTAINABILITY

Specifications

Description 5-IAF (5-iodoacetamido-fluorescein), 25mg, Yellow to Orange, Powder, FITC Dye, 515.26±3 Molecular Weight, 494/518nm Excitation/Emission Wavelength, 63368-54-7 CAS Number, –20°C Storage Temperature, Used for Protein Biology, Protein Labeli
Format Powder
For Use With (Application) Protein labeling and crosslinking, protein biology
Quantity 25 mg
Product Type 5-IAF (5-iodoacetamido-fluorescein)
Molecular Weight (g/mol) 515.26±3
Purity >99% by HPLC
Content And Storage -20°C
Label or Dye FITC (Fluorescein)
Color Yellow to Orange
Emission 518
Excitation 494
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.