missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxycytidine, 99%

Product Code. 15453959
Click to view available options
Quantity:
1 g
2 g
Unit Size:
1g
2g
This item is not returnable. View return policy

Product Code. 15453959

Brand: Thermo Scientific Alfa Aesar J61356.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-53-0 | C9H12IN3O4 | 353.12 g/mol

5-Iodo-2'-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2?-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4,5:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-2′-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2′-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4′,5′:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-53-0
Molecular Formula C9H12IN3O4
Molecular Weight (g/mol) 353.12
MDL Number MFCD00038063
InChI Key WEVJJMPVVFNAHZ-RRKCRQDMSA-N
Synonym 5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
PubChem CID 65050
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Melting Point 180°C to 183°C
Color White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 353.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Iodo-2'-deoxycytidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.