missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxyuridine, ∼99%, MP Biomedicals™

Product Code. 11483040 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483040 25 g 25g
11463040 1 g 1g
11473040 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483040 Supplier MP Biomedicals Supplier No. 0210035725
  • ONLINE EXCLUSIVE PRICE
    1635.00 €
    1548.75 € / 25g
    Save 86.25 €
    5% Off
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-Iodo-2'-deoxyuridine (5-IUdR; IdUrd) is a pyrimidine analog (halogenated thymidine) that has antiviral activity agains vaccinia virus (orthopoxviruses); IdUrd is used as a radiosensitizer in human cancer studies

Chemical Identifiers

CAS 54-42-2
Molecular Formula C9H11IN2O5
Molecular Weight (g/mol) 354.10
MDL Number MFCD00134656
InChI Key XQFRJNBWHJMXHO-RRKCRQDMSA-N
Synonym idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
PubChem CID 5905
ChEBI CHEBI:147675
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

Specifications

Color White
Assay Percent Range ∼99%
Quantity 25 g
Formula Weight 354.1
Physical Form Powder
Chemical Name or Material 5-Iodo-2'-Deoxyuridine, For HPLC analysis
compliance-icons compliance-icons
Product Identifier
  • 5-Iodo-2'-deoxyuridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1A
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-Iodo-2'-deoxyuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.