missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-Deoxyuridine, MP Biomedicals™

Product Code. 11453040 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453040 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453040 Supplier MP Biomedicals Supplier No. 02100357.2
  • ONLINE EXCLUSIVE PRICE
    52.05 €
    49.35 € / 250mg
    Save 2.70 €
    5% Off
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Active against vaccinia virus (orthopoxviruses). 5-Iodo-2'-Deoxyuridine, MP Biomedicals is a pyrimidine analog (halogenated thymidine) that compete with thymidine for incorporation into DNA of replicating cells. This incorporation results in radio sensiti

Specifications

CAS 54-42-2
Melting Point 196°C to 197°C (literature)
Density 2.148g/cm3 (Lit.)
Molecular Formula C9H11IN2O5
MDL Number MFCD00134656
Quantity 250 mg
Synonym idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
Solubility Information Solubility at 25°C in mg/mL: 2.0 in water; 2.0 in 0.2N HCl; 74.0 in 0.2N NaOH; 4.4 in methanol; 2.6 in alc; 0.014 in ether; 0.003 in chloroform; 1.6 in acetone; 1.8 in ethyl acetate; 5.7 in dioxane.
InChI Key XQFRJNBWHJMXHO-RRKCRQDMSA-N
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
Molecular Weight (g/mol) 354.10
PubChem CID 5905
ChEBI CHEBI:147675
Formula Weight 354.1
Percent Purity ∼99% by HPLC
Physical Form Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Iodo-2'-deoxyuridine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1A
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-Iodo-2'-deoxyuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.