missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-phenoxypyridine, ≥95%, Thermo Scientific™

Product Code. 10503002
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10503002 10 g 10g
10304282 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10503002 Supplier Thermo Scientific Maybridge Supplier No. CC19611EA
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 352018-92-9
Molecular Formula C11H8INO
Molecular Weight (g/mol) 297.095
MDL Number MFCD02681952
InChI Key JCEXQQNAXWYXRX-UHFFFAOYSA-N
Synonym 5-iodo-2-phenoxy-pyridine, pyridine,5-iodo-2-phenoxy
PubChem CID 2776503
IUPAC Name 5-iodo-2-phenoxypyridine
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)I

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 297.09
Percent Purity 95%
Chemical Name or Material 5-Iodo-2-phenoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.