missing translation for 'onlineSavingsMsg'
Learn More

5-Iodoindole, 95%

Product Code. 11996721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11996721 25 g 25g
11986721 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11996721 Supplier Thermo Scientific Acros Supplier No. 441590250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16066-91-4 | C8H6IN | 243.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16066-91-4
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.04
InChI Key TVQLYTUWUQMGMP-UHFFFAOYSA-N
Synonym 5-iodoindole, 1h-indole, 5-iodo, 5-iodo indole, 5-iodanyl-1h-indole, zlchem 1099, acmc-1brxb, maybridge1_006499
PubChem CID 2782313
IUPAC Name 5-iodo-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1I

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 95°C to 99°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 25 g
Formula Weight 243.04
Percent Purity 95%
Physical Form Crystalline Solid or Solid
Chemical Name or Material 5-Iodoindole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Iodoindole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.