missing translation for 'onlineSavingsMsg'
Learn More

5-Iodoindole, 98%

Product Code. 11459077
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459077 25 g 25g
11449077 5 g 5g
11469077 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459077 Supplier Thermo Scientific Alfa Aesar Supplier No. B20638.14
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16066-91-4 | C8H6IN | 243.047 g/mol

5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16066-91-4
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.047
MDL Number MFCD00220065
InChI Key TVQLYTUWUQMGMP-UHFFFAOYSA-N
Synonym 5-iodoindole, 1h-indole, 5-iodo, 5-iodo indole, 5-iodanyl-1h-indole, zlchem 1099, acmc-1brxb, maybridge1_006499
PubChem CID 2782313
IUPAC Name 5-iodo-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1I

Specifications

Melting Point 97°C to 100°C
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 243.05
Percent Purity 98%
Chemical Name or Material 5-Iodoindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.