missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 5-Iodotubercidin

Product Code. 18054072
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18054072 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18054072 Supplier R&D Systems™ Supplier No. 1745/1
  • ONLINE EXCLUSIVE PRICE
    234.30 €
    210.00 € / 1mg
    Save 24.30 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent adenosine kinase inhibitor; also inhibits nucleoside transporters and a range of other kinases

Potent adenosine kinase inhibitor (IC50 = 26 nM). Also nucleoside transporter inhibitor (IC50 values are < 25 nM, 7 μM and 15 μM for inhibition of [3H]adenosine, [3H]uridine and [3H]formycin B uptake respectively). Strongly stimulates glycogen synthesis in hepatocytes via activation of glycogen synthase. Also inhibits CK1, insulin receptor tyrosine kinase, phosphorylase kinase, PKA, CK2 and PKC (IC50 values are 0.4, 3.5, 5-10, 5-10, 10.9 and 27.7 μM respectively). Decreases hippocampal DNA methylation through adenosine kinase inhibition in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 24386-93-4
Quantity 1 mg
Product Line Tocris
Molecular Formula C11H13IN4O4
Formula Weight 392.15
Purity 97%
Molecular Weight (g/mol) 392.15
Chemical Name or Material 5-Iodotubercidin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.