missing translation for 'onlineSavingsMsg'
Learn More

5-(Isocyanatomethyl)-1,3-benzodioxole, ≥97%, Thermo Scientific™

Product Code. 10247723
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10247723 250 mg 250mg
10600904 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10247723 Supplier Thermo Scientific Maybridge Supplier No. CC50806CB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 71217-46-4
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD08435909
InChI Key RIUNOJGBBOBVDE-UHFFFAOYSA-N
Synonym 5-isocyanatomethyl-2h-1,3-benzodioxole, 5-isocyanatomethyl-1,3-benzodioxole, 1,3-benzodioxole,5-isocyanatomethyl, 1,3-benzodioxole, 5-isocyanatomethyl, 3,4-methylenedioxy benzyl isocyanate, 1,3-benzodioxol-5-ylmethyl isocyanate, 5-isocyanatomethyl benzo d 1,3 dioxole, 2h-benzo 3,4-d 1,3-dioxolan-5-ylmethanisocyanate
PubChem CID 15932354
IUPAC Name 5-(isocyanatomethyl)-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)CN=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-(Isocyanatomethyl)-1, 3-benzodioxole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.