missing translation for 'onlineSavingsMsg'
Learn More

5-Mercapto-3-phenyl-1,3,4-thiadiazole-2(3H)thione potassium salt, Thermo Scientific™

Product Code. 11359728
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359728 2 g 2g
11369728 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359728 Supplier Thermo Scientific Alfa Aesar Supplier No. 044219.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a complexing reagent for Bi, Te, Se and other metal ions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6336-51-2
Molecular Formula C8H5KN2S3
Molecular Weight (g/mol) 264.42
MDL Number MFCD00014092
InChI Key QSKDCVGFAPHSHX-UHFFFAOYSA-M
Synonym 1,3,4-thiadiazolidine-2,5-dithione, 3-phenyl-, potassium salt, 3-phenyl-1,3,4-thiadiazolidine-2,5-dithione potassium, potassium 5-mercapto-3-phenyl-1,3,4-thiadiazole-2-thione, 3-phenyl-5-sulfanyl-1,3,4-thiadiazole-2-thione potassium, 5-mercapto-3-phenyl-1,3,4-thiadiazole-2 3h thione potassium salt
PubChem CID 2775614
IUPAC Name potassium;4-phenyl-5-sulfanylidene-1,3,4-thiadiazole-2-thiolate
SMILES C1=CC=C(C=C1)N2C(=S)SC(=N2)[S-].[K+]

Specifications

Quantity 2 g
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
Formula Weight 264.43
Physical Form Powder
Chemical Name or Material 5-Mercapto-3-phenyl-1,3,4-thiadiazole-2 (3H)thione potassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.