missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-mercaptobenzimidazole, 99+%

Product Code. 10792861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792861 25 g Glass bottle 25g
10784543 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10792861 Supplier Thermo Scientific Acros Supplier No. 275370250
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37052-78-1 | C8H8N2OS | 180.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37052-78-1
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.23
MDL Number MFCD00134581
InChI Key KOFBRZWVWJCLGM-UHFFFAOYSA-N
Synonym 5-methoxy-2-mercaptobenzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-methoxy-1h-benzimidazole, 5-methoxy-1h-benzoimidazole-2-thiol, 1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 5-methoxybenzimidazole-2-thiol
PubChem CID 665603
IUPAC Name 5-methoxy-1,3-dihydrobenzimidazole-2-thione
SMILES COC1=CC=C2NC(=S)NC2=C1

Specifications

Absorbance (1% soln. in methanol) (1 cm cell) no other absorption observed from 360 to 600nm, (1% soln. in methanol) (1 cm cell) no other absorption observed from 360 to 600nm at 0nm, 0.4 max. at 360nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point 255°C to 262°C
Color Brown-Yellow to White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in sodium hydroxide solution
Formula Weight 180.23
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 5-Methoxy-2-mercaptobenzimidazole, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.