missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methyl-3-indoleacetic acid, 98%

Product Code. 10183620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183620 1 g Glass bottle 1g
10367120 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10183620 Supplier Thermo Scientific Acros Supplier No. 147510010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2882-15-7 | C12H13NO3 | 219.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2882-15-7
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00005618
InChI Key TXWGINUZLBAKDF-UHFFFAOYSA-N
Synonym 5-methoxy-2-methyl-3-indoleacetic acid, 5 miaa, 2-5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 5-methoxy-2-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 5-methoxy-2-methyl, n-deschlorobenzoyl indomethacin, 5-methoxy-2-methylindole-3-acetic acid, 5-methoxy-methylindoleacetic acid, deschlorobenzoyl indomethacin, 5-methoxy-2-methyl indole acetic acid
PubChem CID 76151
IUPAC Name 2-(5-methoxy-2-methyl-1H-indol-3-yl)acetic acid
SMILES CC1=C(C2=C(N1)C=CC(=C2)OC)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 162°C to 165°C
Color Beige or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 219.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxy-2-methyl-3-indoleacetic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.