missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxy-2-(trifluoromethyl)pyridine, Thermo Scientific™
Specifications
Specifications
| CAS | 216766-13-1 |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C7H6F3NO |
| MDL Number | MFCD09864699 |
| Quantity | 5 g |
| Synonym | 5-methoxy-2-trifluoromethyl pyridine, 5-methoxy-2-trifluoromethyl-pyridine, pubchem23560, 3-methoxy-6-trifluoromethylpyridine, 3-methoxy-6-trifluoromethyl pyridine, pyridine,5-methoxy-2-trifluoromethyl |
| InChI Key | MRHJZBSCOLVMGA-UHFFFAOYSA-N |
| SMILES | COC1=CN=C(C=C1)C(F)(F)F |
| IUPAC Name | 5-methoxy-2-(trifluoromethyl)pyridine |
| Molecular Weight (g/mol) | 177.13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?