missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-7-methyl-1H-indole, 95%, BLD Pharm™

Product Code. 31112014
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31112014 250 mg
31109465 5 g
31102295 1 g
31110121 10 g
31103061 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31112014 Supplier BLD Pharm Supplier No. BD104527250mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 61019-05-4
Molecular Formula C10H11NO
MDL Number MFCD09027022
Quantity 250 mg
SMILES COC1=CC2=C(NC=C2)C(C)=C1
Molecular Weight (g/mol) 161.20
Percent Purity 95%
Chemical Name or Material 5-Methoxy-7-methyl-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.