missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxybenzofuroxan, 97%, Thermo Scientific™

Product Code. 11494904
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494904 25 g 25g
11484904 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494904 Supplier Thermo Scientific Alfa Aesar Supplier No. A13822.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-49-3
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00068052
InChI Key ABSKHUSUHODMEL-UHFFFAOYSA-N
Synonym 5-methoxybenzofurazan-1-oxide, 5-methoxy-2,1,3-benzoxadiazole 1-oxide, 5-methoxy-2,1,3-benzoxadiazol-1-ium-1-olate, 5-methoxybenzofuroxane, 5-methoxybenzofurazane 1-oxide, 5-methoxybenzofurazan, 1-oxide, 5-methoxybenzo c 1,2,5-oxadiazol-1-ol, 2,1,3-benzoxadiazole,5-methoxy-,1-oxide, 2,1,3-benzoxadiazole, 5-methoxy-, monooxide
PubChem CID 321028
IUPAC Name 5-methoxy-1-oxido-2,1,3-benzoxadiazol-1-ium
SMILES COC1=CC2=NO[N+](=C2C=C1)[O-]

Specifications

Melting Point 112°C to 114°C
Quantity 25 g
Beilstein 643053
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 5-Methoxybenzofuroxan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.