missing translation for 'onlineSavingsMsg'
Learn More

5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 11946111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11946111 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11946111 Supplier Thermo Scientific Acros Supplier No. 445110010
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 882679-40-5
Melting Point 157.0°C to 159.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Molecular Formula C15H21BO4
MDL Number MFCD11520530
Quantity 1 g
Synonym methyl 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 5-methoxycarbonyl-2-methylphenylboronic acid pinacol ester, amtb518, 5-methoxycarbonyl-2-methylphenylboronic acid, pinacol ester, methyl 4-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid methyl ester, benzoicacid, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, methyl4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid methyl ester
InChI Key HAPIXNBOBZHNCA-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(B2OC(C)(C)C(C)(C)O2)=C(C)C=C1
IUPAC Name methyl 4-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol) 276.14
PubChem CID 53217136
Formula Weight 276.14
Percent Purity 95%
Chemical Name or Material 5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • 5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.