missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-carboxaldehyde, 99+%, Thermo Scientific™

Product Code. 10229223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
100mg
1g
5g
Product Code. Quantity Packaging unitSize
10229223 100 mg Glass Bottle 100mg
10685362 1 g Glass Bottle 1g
10595881 5 g Glass Bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10229223

Brand: Thermo Scientific Acros 125891000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10601-19-1
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005623
InChI Key TUWARWGEOHQXCO-UHFFFAOYSA-N
Synonym 5-methoxyindole-3-carboxaldehyde, 3-formyl-5-methoxyindole, 5-methoxyindole-3-carboxyaldehyde, 5-methoxyindole-3-carbaldehyde, 5-methoxyindole-3-aldehyde, 5-methoxy-3-formylindole, 1h-indole-3-carboxaldehyde, 5-methoxy, 5-methoxy-indol-3-aldehyde, unii-63c50zw2d0, 5-methoxy-1h-indole-3-carboxaldehyde
PubChem CID 82758
IUPAC Name 5-methoxy-1H-indole-3-carbaldehyde
SMILES COC1=CC=C2NC=C(C=O)C2=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 180.0°C to 184.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Quantity 100 mg
Beilstein 21, II, 424
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 175.19
Percent Purity ≥99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 5-Methoxyindole-3-carboxaldehyde, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxyindole-3-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.