missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypiperonal, 97%

Product Code. 11388035
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388035 25 g 25g
11368035 1 g 1g
11378035 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388035 Supplier Thermo Scientific Alfa Aesar Supplier No. L08213.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5780-07-4 | C9H8O4 | 180.16 g/mol

5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents. Store under dry inert gas. Store away from air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5780-07-4
MDL Number MFCD00016900

Specifications

Melting Point 126°C to 132°C
Molecular Formula C9H8O4
Quantity 25 g
Beilstein 384077
Sensitivity Air Sensitive
Synonym myristicin aldehyde, 3-methoxy-4,5-methylenedioxybenzaldehyde, 3-methoxypiperonal, 1,3-benzodioxole-5-carboxaldehyde, 7-methoxy, 5-methoxypiperonal, 7-methoxy-benzo 1,3 dioxole-5-carbaldehyde, 7-methoxybenzo d 1,3 dioxole-5-carbaldehyde, 7-methoxy-2h-1,3-benzodioxole-5-carbaldehyde, 7-methoxybenzo-1,3-dioxole-5-carboxaldehyde, 7-methoxybenzo 1,3 dioxole-5-carbaldehyde
Solubility Information Insoluble in water.
InChI Key LOFRBHZYZCIOJO-UHFFFAOYSA-N
SMILES COC1=CC(=CC2=C1OCO2)C=O
IUPAC Name 7-methoxy-1,3-benzodioxole-5-carbaldehyde
Molecular Weight (g/mol) 180.16
PubChem CID 22016
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 5-Methoxypiperonal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.