missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-benzothiophen-2-ylboronic acid, 97%, Thermo Scientific™

Product Code. 10671245
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10671245 250 mg Amber Glass Bottle 250mg
10472223 1 g Amber Glass Bottle 1g
10205964 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10671245 Supplier Thermo Scientific Maybridge Supplier No. MO00912CB
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 136099-65-5
Molecular Formula C9H9BO2S
Molecular Weight (g/mol) 192.04
MDL Number MFCD06200866
InChI Key DHNHZPQPQAINDI-UHFFFAOYSA-N
Synonym 5-methylbenzo b thiophene-2-boronic acid, 5-methylbenzothiphene-2-boronic acid, 5-methylbenzo b thiophen-2-ylboronic acid, 5-methyl-1-benzothiophen-2-yl boronic acid, 5-methylbenzothiophene-2-boronic acid, 2-borono-5-methylbenzo b thiophene, 5-methyl-benzo b thiophen-2-ylboronic acid, 5-methylbenzothiophen-2-yl boronic acid, d00wzi, 5-methylbenzo b thiophen-2-yl boronic acid
PubChem CID 2794641
IUPAC Name (5-methyl-1-benzothiophen-2-yl)boronic acid
SMILES CC1=CC=C2SC(=CC2=C1)B(O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 192.05
Percent Purity 97%
Chemical Name or Material 5-Methylbenzo[b]thiophene-2-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.