missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-benzothiophene-2-sulfonyl chloride, Thermo Scientific™

Product Code. 10340883
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10340883 250 mg Amber Glass Bottle 250mg
10294762 1 g Amber Glass Bottle 1g
10356754 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10340883 Supplier Thermo Scientific Maybridge Supplier No. MO07074CB
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 90273-30-6
Molecular Formula C9H7ClO2S2
Molecular Weight (g/mol) 246.72
MDL Number MFCD06200880
InChI Key YGGJYXHTCDQVDB-UHFFFAOYSA-N
Synonym 5-methylbenzothiophene-2-sulfonyl chloride, 5-methylbenzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 5-methyl, 5-methyl-1-benzothiophene-2-sulfonylchloride, 5-methyl-benzo b thiophenesulfonyl chloride, 5-methyl-benzothiophene-2-sulfonyl chloride, chloro 5-methylbenzo b thiophen-2-yl sulfone, benzo b thiophene-2-sulfonylchloride, 5-methyl
PubChem CID 2794771
IUPAC Name 5-methyl-1-benzothiophene-2-sulfonyl chloride
SMILES CC1=CC=C2SC(=CC2=C1)S(Cl)(=O)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 246.74
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-benzothiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.