missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-hexyne, 98%

Product Code. 11372326
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11372326 5 g 5g
11382326 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11372326

Brand: Thermo Scientific Alfa Aesar L06014.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2203-80-7 | C7H12 | 96.173 g/mol

5-Methyl-1-hexyne is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1-hexyne is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2203-80-7
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00041610
InChI Key HKNANEMUCJGPMS-UHFFFAOYSA-N
Synonym 5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa
PubChem CID 137490
IUPAC Name 5-methylhex-1-yne
SMILES CC(C)CCC#C

Specifications

Density 0.732
Boiling Point 91°C to 93°C
Flash Point −4°C (24°F)
Refractive Index 1.4075
Quantity 5 g
UN Number UN3295
Beilstein 1731845
Solubility Information Not miscible in water.
Formula Weight 96.17
Percent Purity 98%
Chemical Name or Material 5-Methyl-1-hexyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.