missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11363256
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363256 250 mg 250mg
11343256 1 g 1g
11353256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363256 Supplier Thermo Scientific Alfa Aesar Supplier No. L12594.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91138-00-0
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD00067831
InChI Key USSMIQWDLWJQDQ-UHFFFAOYSA-N
Synonym 5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
PubChem CID 145221
IUPAC Name 5-methyl-1-phenylpyrazole-4-carboxylic acid
SMILES CC1=C(C=NN1C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 165°C to 169°C
Quantity 250 mg
Beilstein 162776
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.