missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10417612
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10417612 1 g 1g
10382055 5 g 5g
10702017 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10417612 Supplier Thermo Scientific Maybridge Supplier No. CC15501DA
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 91138-00-0
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
InChI Key USSMIQWDLWJQDQ-UHFFFAOYSA-N
Synonym 5-methyl-1-phenyl-1h-pyrazole-4-carboxylic acid, 3-methyl-2-phenylpyrazole-4-carboxylic acid, 3-methyl-2-phenyl-4-pyrazolecarboxylic acid, akos pao-0331, acmc-20am2c, rarechem al be 1317, maybridge1_002413, buttpark 33\08-16, cbi-bb zero/006129, 5-methyl-1-phenyl-1h-pyrazole-4-carboxylicacid
PubChem CID 145221
IUPAC Name 5-methyl-1-phenylpyrazole-4-carboxylic acid
SMILES CC1=C(C=NN1C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.