missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, 95+%, Thermo Scientific™

Product Code. 11350577
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11350577 5 g 5g
10468182 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350577 Supplier Thermo Scientific Maybridge Supplier No. CC15503DE
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 342405-38-3
Molecular Formula C10H9ClN2O2S
Molecular Weight (g/mol) 256.70
MDL Number MFCD02681919
InChI Key IRJCRNZTDRODFQ-UHFFFAOYSA-N
Synonym 5-methyl-1-phenyl-1h-pyrazole-4-sulfonyl chloride, 5-methyl-1-phenyl-1h-pyrazole-4-sulphonyl chloride, 5-methyl-1-phenyl-pyrazole-4-sulfonyl chloride, 4-chlorosulphonyl-1-phenyl-1h-pyrazole, 5-methyl-1-phenyl-4-pyrazolesulfonyl chloride, 5-methyl-1-phenylpyrazol-4-sulfonyl chloride, chloro 5-methyl-1-phenylpyrazol-4-yl sulfone, 1h-pyrazole-4-sulfonylchloride, 5-methyl-1-phenyl, 4-chlorosulphonyl-5-methyl-1h-pyrazol-1-yl benzene
PubChem CID 2776435
IUPAC Name 5-methyl-1-phenylpyrazole-4-sulfonyl chloride
SMILES CC1=C(C=NN1C1=CC=CC=C1)S(Cl)(=O)=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 256.71
Percent Purity 95%
Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.