missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-benzotriazole, 98+%

Product Code. 11463314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463314 500 g 500g
11443314 25 g 25g
11453314 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463314 Supplier Thermo Scientific Alfa Aesar Supplier No. A10615.36
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 136-85-6 | C7H7N3 | 133.15 g/mol

5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-85-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005702
InChI Key LRUDIIUSNGCQKF-UHFFFAOYSA-N
Synonym 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
PubChem CID 8705
ChEBI CHEBI:83455
IUPAC Name 5-methyl-2H-benzotriazole
SMILES CC1=CC2=NNN=C2C=C1

Specifications

Melting Point 77°C to 84°C
pH 5.3
Boiling Point 210°C to 212°C (12 mmHg)
Odor Odorless
Quantity 500 g
Beilstein 116658
Solubility Information Soluble in chloroform,methanol,water (6.0g/L at 25°C),ethanol,and toluene (slightly).
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material 5-Methyl-1H-benzotriazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.