missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10495712
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10495712 1 g Amber Glass Bottle 1g
10565184 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10495712 Supplier Thermo Scientific Maybridge Supplier No. SPB07714DA
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 402-61-9
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00090754
InChI Key WSMQKESQZFQMFW-UHFFFAOYSA-N
Synonym 3-methyl-1h-pyrazole-5-carboxylic acid, 5-methylpyrazole-3-carboxylic acid, 5-methyl-2h-pyrazole-3-carboxylic acid, 3-methylpyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-methyl, 5-methylpyrazole-3-carboxylicacid, 3-methyl-1h-pyrazole-5-carboxylicacid, pyrazole-3-carboxylic acid, 5-methyl, pubchem7842, 5-methyl-2h-pyrazole-3-carboxylicacid
PubChem CID 9822
ChEBI CHEBI:74739
IUPAC Name 5-methyl-1H-pyrazole-3-carboxylic acid
SMILES CC1=CC(=NN1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Merck Index 15, 6188
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 5-Methyl-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.