missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2'-deoxycytidine, 99%

Product Code. 15446789
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446789 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446789 Supplier Thermo Scientific Alfa Aesar Supplier No. J65209.03
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 838-07-3 | C10H15N3O4 | 241.247 g/mol

5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-2′-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditio
TRUSTED_SUSTAINABILITY

Specifications

CAS 838-07-3
Color White
Molecular Formula C10H15N3O4
MDL Number MFCD00006549
Quantity 1 g
Synonym 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference
Solubility Information Soluble in water.
InChI Key LUCHPKXVUGJYGU-XLPZGREQSA-N
SMILES CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
Molecular Weight (g/mol) 241.247
PubChem CID 440055
ChEBI CHEBI:47876
Formula Weight 241.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Methyl-2'-deoxycytidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.