missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furaldehyde, 98%

Product Code. 11490803
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490803 25 g 25g
11400813 100 g 100g
11410813 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490803 Supplier Thermo Scientific Alfa Aesar Supplier No. A13264.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-02-0 | C6H6O2 | 110.112 g/mol

5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

Solubility
Soluble in alcohol and water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Density 1.106
Boiling Point 186°C to 188°C
Flash Point 72°C (161°F)
Refractive Index 1.531
Quantity 25 g
Beilstein 106895
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol and water.
Formula Weight 110.11
Percent Purity 98%
Chemical Name or Material 5-Methyl-2-furaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.