missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hex-2-enoic Acid, TRC

Product Code. 30464483
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464483 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464483 Supplier Toronto Research Chemicals Supplier No. TRCM31196050MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methyl-2-hex-2-enoic Acid
CAS 41653-96-7
Molecular Formula C7 H12 O2
Quantity 50 mg
InChI Formula InChI=1S/C7H12O2/c1-6(2)4-3-5-7(8)9/h3,5-6H,4H2,1-2H3,(H,8,9)/b5-3+
SMILES CC(C)C\C=C\C(=O)O
IUPAC Name (E)-5-methylhex-2-enoic acid
Molecular Weight (g/mol) 128.17
Formula Weight 128.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.