missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hexanone, 99%, Honeywell™

Product Code. 15635790 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
20 L
Packaging:
Glass bottle
Unit Size:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15635790 1 L Glass bottle 1 L
15645790 20 L Glass bottle 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15635790 Supplier Honeywell Research Chemicals Supplier No. 5377051L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C

Specifications

Density 0.814g/mL at 25°C
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2COCH3
Quantity 1 L
UN Number UN2302
Beilstein 506163
Formula Weight 114.19g/mol
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.