missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hexanone, 99%

Product Code. 11388526
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388526 100 mL 100mL
11398526 500 mL 500mL
11308536 2500 mL 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388526 Supplier Thermo Scientific Alfa Aesar Supplier No. L17584.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-12-3 | C7H14O | 114.188 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (5.4 g/L)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C

Specifications

Melting Point -74°C
Density 0.812
Boiling Point 144°C to 145°C
Flash Point 41°C (105°F)
Refractive Index 1.4075
Quantity 100 mL
UN Number UN2302
Beilstein 506163
Solubility Information Soluble in water. (5.4g/L)
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hexanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.