missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hexanone, 99%

Product Code. 10173720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173720 1 L Glass bottle 1 L
10796361 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173720 Supplier Thermo Scientific Acros Supplier No. 149660010
  • ONLINE EXCLUSIVE PRICE
    77.15 €
    74.90 € / 1 L
    Save 2.25 €
    2% Off
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 110-12-3 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point -74.0°C
Color Colorless
Density 0.8100g/mL
Boiling Point 145.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2COCH3
Refractive Index 1.4060 to 1.4080
Quantity 1 L
Specific Gravity 0.81
Solubility Information Solubility in water: 5.4g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 114.19
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-hexanone, 99%
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 5-Methyl-2-hexanone
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Indicaciones de peligro
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.