missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-2-phenyl-2H-1,2,3-triazol-4-yl)methanol, 95%, Thermo Scientific™

Product Code. 10283012
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10283012 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10283012 Supplier Thermo Scientific Maybridge Supplier No. CC16209DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 13322-19-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C10H11N3O
MDL Number MFCD02682048
Quantity 1 g
Synonym 5-methyl-2-phenyl-2h-1,2,3-triazol-4-yl methanol, 2h-1,2,3-triazole-4-methanol, 5-methyl-2-phenyl, 5-methyl-2-phenyl-1,2,3-triazol-4-yl methanol, 5-methyl-2-phenyl-1,2,3-triazol-4-yl methan-1-ol, 2-phenyl-4-hydroxymethyl-5-methyl-2h-1,2,3-triazole, 5-methyl-2-phenyltriazol-4-yl methanol, 2h-1,2,3-triazole-4-methanol,5-methyl-2-phenyl, 5-methyl-2-phenyl-1,2,3-triazol4-yl methan-1-ol, 5-methyl-2-phenyl-2h-1,2,3 triazol-4-yl-methanol, 5-methyl-2-phenyl-2h-1,2,3-triazol-4-yl methanol #
InChI Key VYHJVMFMRIGUFV-UHFFFAOYSA-N
SMILES CC1=NN(N=C1CO)C1=CC=CC=C1
IUPAC Name (5-methyl-2-phenyl-2H-1,2,3-triazol-4-yl)methanol
Molecular Weight (g/mol) 189.22
PubChem CID 563195
Formula Weight 189.22
Percent Purity 95%
Chemical Name or Material (5-Methyl-2-phenyl-2H-1, 2, 3-triazol-4-yl)methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.