missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-2-phenyl-3-furyl)methylamine, 97%, Thermo Scientific™

Product Code. 10286333
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10286333 250 mg Amber Glass Bottle 250mg
10265804 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10286333 Supplier Thermo Scientific Maybridge Supplier No. CC49713CB
 Restricted Item
Estimated Shipment: 12-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 771572-29-3
Molecular Formula C12H13NO
Molecular Weight (g/mol) 187.24
MDL Number MFCD06213531
InChI Key PQZVRVFUCJMCRZ-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-3-furyl methylamine, 5-methyl-2-phenylfuran-3-yl methanamine, 1-5-methyl-2-phenylfuran-3-yl methanamine, 3-furanmethanamine,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methanamine, 5-methyl-2-phenylfur-3-yl methylamine
PubChem CID 24229505
IUPAC Name (5-methyl-2-phenylfuran-3-yl)methanamine
SMILES CC1=CC(CN)=C(O1)C1=CC=CC=C1

Tekniske data

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material (5-Methyl-2-phenyl-3-furyl)methylamine
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.