missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-thiophenecarboxaldehyde, 98%

Product Code. 10467900 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10467900

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13679-70-4 | C6H6OS | 126.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13679-70-4
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.18
MDL Number MFCD00005434
InChI Key VAUMDUIUEPIGHM-UHFFFAOYSA-N
Synonym 5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 2-formyl-5-methylthiophene, 5-methyl-2-formylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methylthiophene-2-aldehyde, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehyde
PubChem CID 61663
IUPAC Name 5-methylthiophene-2-carbaldehyde
SMILES CC1=CC=C(S1)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.1800g/mL
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5800 to 1.584
Quantity 10 mL
Beilstein 17, I, 151
Specific Gravity 1.18
Formula Weight 126.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-thiophenecarboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.