missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+%

Product Code. 15468117
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468117 250 mg 250mg
15458117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468117 Supplier Thermo Scientific Alfa Aesar Supplier No. H51531.MD
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 848613-91-2 | C16H31NSSn | 388.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 848613-91-2
Molecular Formula C16H31NSSn
Molecular Weight (g/mol) 388.201
MDL Number MFCD09025809
InChI Key LALGELPHLJBAEK-UHFFFAOYSA-N
Synonym 5-methyl-2-tributylstannyl thiazole, 5-methyl-2-tributylstannyl-1,3-thiazole, 5-methyl-2-tri-n-butylstannyl thiazole, acmc-209px3, 5-methyl-2-tributylstannyl-thiazole, thiazole, 5-methyl-2-tributylstannyl, 5-methyl-2-tributylstannanyl-thiazole, 5-methyl-2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl-5-methylthiazole
PubChem CID 16427131
IUPAC Name tributyl-(5-methyl-1,3-thiazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=C(S1)C

Specifications

Quantity 250 mg
UN Number UN2788
Formula Weight 388.19
Percent Purity ≥90%
Chemical Name or Material 5-Methyl-2-(tri-n-butylstannyl)thiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.