missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11310557
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11310557 10 g Amber Glass Bottle 10g
10079161 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310557 Supplier Thermo Scientific Maybridge Supplier No. CC00703EA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-02-4
Molecular Formula C6H4ClF3O3S
Molecular Weight (g/mol) 248.6
InChI Key GSYXWDYWBRICSD-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl furan-3-sulfonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonyl chloride, 3-furansulfonylchloride, 5-methyl-2-trifluoromethyl, 5-methyl-2-trifluoromethyl furan-3-sulphonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonylchloride, 5-methyl-2-trifluoromethyl-furan-3-sulfonyl chloride, chloro 5-methyl-2-trifluoromethyl 3-furyl sulfone, 3-furansulfonyl chloride, 5-methyl-2-trifluoromethyl-9ci
PubChem CID 2779895
IUPAC Name 5-methyl-2-(trifluoromethyl)furan-3-sulfonyl chloride
SMILES CC1=CC(=C(O1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.