missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-methylene-2-hexanone, TRC

Product Code. 30475415
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475415 100 mg 100mg
30470689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475415 Supplier Toronto Research Chemicals Supplier No. TRCM322950100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187-87-7
Molecular Formula C8 H14 O
Molecular Weight (g/mol) 126.2
Synonym 4-Methyl-3-methylene-2-hexanone
IUPAC Name 5-methyl-3-methylidenehexan-2-one
SMILES CC(C)CC(=C)C(=O)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H14O/c1-6(2)5-7(3)8(4)9/h6H,3,5H2,1-2,4H3
Formula Weight 126.1045
Chemical Name or Material 5-Methyl-3-methylene-2-hexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.