missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenylisoxazole-4-carbonyl chloride, 90%, Thermo Scientific™

Product Code. 10374372
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10374372 1 g 1g
10782093 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374372 Supplier Thermo Scientific Maybridge Supplier No. CC00502DA
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 16883-16-2
Molecular Formula C11H8ClNO2
Molecular Weight (g/mol) 221.64
MDL Number MFCD00052205
InChI Key HXEVQMXCHCDPSO-UHFFFAOYSA-N
Synonym 5-methyl-3-phenylisoxazole-4-carbonyl chloride, pmic chloride, unii-97ot2t1w6v, 3-phenyl-5-methylisoxazole-4-carbonyl chloride, 4-isoxazolecarbonyl chloride, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarbonyl chloride, 3-phenyl-5-methyl-isoxazole-4-carbonyl, pubchem8723, acmc-20am25, dsstox_cid_31452
PubChem CID 85624
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Density 1.2700g/mL
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.64
Percent Purity 90%
Chemical Name or Material 5-Methyl-3-phenylisoxazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.