missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%

Product Code. 15460817
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460817 25 g 25g
15470817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460817 Supplier Thermo Scientific Alfa Aesar Supplier No. L01142.14
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1136-45-4 | C11H9NO3 | 203.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1136-45-4
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
MDL Number MFCD00003153
InChI Key PENHKTNQUJMHIR-UHFFFAOYSA-N
Synonym 5-methyl-3-phenylisoxazole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazolecarboxylic acid, 3-phenyl-5-methylisoxazol-4-carbonsaeure, 3-phenyl-5-methylisoxazole-4-carboxylic acid, isoxazole-4-carboxylic acid, 5-methyl-3-phenyl, 5-methyl-3-phenyl-isoxazole-4-carboxylic acid, 5-methyl-3-phenylisoxazole-4-carboxylicacid, 3-phenyl-5-methylisoxazol-4-carbonsaeure german, 5-methyl-3-phenyl-4-isooxazolic acid
PubChem CID 14343
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 191°C to 193°C
Odor Odorless
Quantity 25 g
Beilstein 164939
Formula Weight 203.2
Percent Purity 99%
Chemical Name or Material 5-Methyl-3-phenylisoxazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.