missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-4-isoxazolesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 10541512
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10541512 10 g Amber Glass Bottle 10g
10436652 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10541512 Supplier Thermo Scientific Maybridge Supplier No. CC00271EA
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 321309-26-6
Molecular Formula C4H4ClNO3S
Molecular Weight (g/mol) 181.59
MDL Number MFCD02681975
InChI Key STYUNYVEEFPQMF-UHFFFAOYSA-N
Synonym 5-methyl-4-isoxazolesulfonyl chloride, 5-methylisoxazole-4-sulfonyl chloride, 4-isoxazolesulfonyl chloride, 5-methyl, 4-chlorosulfonyl-5-methylisoxazole, chloro 5-methylisoxazol-4-yl sulfone, 4-chlorosulphonyl-5-methylisoxazole, 4-isoxazolesulfonylchloride, 5-methyl, 5-methyl-4-isoxazole sulfonyl chloride, 5-methyl-isoxazole-4-sulfonyl chloride, 5-methylisoxazole-4-sulphonyl chloride
PubChem CID 2776141
IUPAC Name 5-methyl-1,2-oxazole-4-sulfonyl chloride
SMILES CC1=C(C=NO1)S(=O)(=O)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 181.6
Percent Purity 95%
Chemical Name or Material 5-Methyl-4-isoxazolesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.