missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-4-nitroimidazole, TRC

Product Code. 30479411
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479411 5 g 5g
30470356 25 g 25g
30470355 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479411 Supplier Toronto Research Chemicals Supplier No. TRCM3238105G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14003-66-8
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.1
Synonym 4-Methyl-5-nitroimidazole, NSC 156634, RP 8549
IUPAC Name 4-methyl-5-nitro-1H-imidazole
SMILES Cc1nc[nH]c1[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H5N3O2/c1-3-4(7(8)9)6-2-5-3/h2H,1H3,(H,5,6)
Formula Weight 127.0382
Chemical Name or Material 5-Methyl-4-nitroimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.