missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzotriazole, TRC

Product Code. 30135260
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135260 10 g 10g
17974655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135260 Supplier Toronto Research Chemicals Supplier No. TRCM28878510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-85-6
InChI Formula InChI=1S/C7H7N3/c1-5-2-3-6-7(4-5)9-10-8-6/h2-4H,1H3,(H,8,9,10)
IUPAC Name 5-methyl-1H-benzotriazole
Molecular Formula C7 H7 N3
Molecular Weight (g/mol) 133.15
Chemical Name or Material 5-Methyl-1H-benzotriazole

Specifications

Quantity 10 g
Synonym 1H-Benzotriazole, 5-methyl- (8CI,9CI), Benzotriazole, 5-methyl- (6CI,7CI), 6-Methyl-1H-benzotriazole, 5-Methyl-1,2,3-benzotriazole, 5-Methyl-1H-benzo[d][1,2,3]triazole, 5-Methyl-1H-benzotriazole, 5-Methylbenzotriazole, 5-Tolyltriazole, 5M-BTA, 5MBT, 6-Methyl-1,2,3-benzotriazole, 6-Tolyltriazole, NSC 122012, TTA, Tolutriazole, Verzone VT 120M
SMILES Cc1ccc2[nH]nnc2c1
Formula Weight 133.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.